Skip to Content
Merck
All Photos(1)

Documents

R8812N-10

Sigma-Aldrich

Rat Striatal Neurons: RStN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat forebrain striatum

Quality Level

packaging

pkg of 1,000,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

Adherent. When cultured under the recommended conditions, cells arborize and form complex neurite network in one week.

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Rat Striatal Neurons (RStN) are derived from striatum of normal embryonic rat (gestation day 18). >1,000,000 cells cryopreserved in Serum-free Freezing Medium.

Cell Line Origin

Forebrain

Application

Basic research on input signal processing, interactions among cholinergic, dopaminergic, and GABAergic neurotransmitter systems, voluntary movement, reward-based habit learning, addiction, procedural memory formation, cognitive dysfunction, Parkinson′s disease, disease mechanisms, pathophysiologies, drug screening and development, toxicity testing, immunostaining, live cell imaging, co-culturing, electrophysiology.

Components

Serum-free Freezing Medium

Quality

Each lot was tested for proper morphology, Stain positive for β III-Tubulin, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service