Skip to Content
Merck
All Photos(1)

Key Documents

R7397

Sigma-Aldrich

ProtectRNA RNase Inhibitor 500× Concentrate

RNase inhibitor for in situ hybridization assays

Synonym(s):

Ribonuclease inhibitor ProtectRNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.52

Quality Level

form

liquid

storage temp.

2-8°C

General description

Not recommended in systems where other enzymatic activity is required.

ProtectRNA ribonuclease(RNase) Inhibitor inhibits most nucleic acid binding enzymes such as RNase and DNase, reverse transcriptase, some RNA and DNA polymerases, protein synthesis enzymes, and several restriction endonucleases. In contrast, ProtectRNA does not interfere with DNA and RNA hybridizations. The action ofProtectRNA as a nuclease inhibitor can be used as an advantage for in situ hybridization. ProtectRNA is useful during all steps of in situ hybridization for tissue RNA protection from RNase degradation. It is recommended to add ProtectRNA to all aqueous solutions used, from as early as the deparaffinization step until the hybridization is completed.

Application

ProtectRNA RNase Inhibitor 500× Concentrate has been used in:
  • RNA extraction
  • tissue sectioning and staining for laser capture microscopy
  • the isolation of blood vessels from hippocampi
ProtectRNA RNase Inhibitor 500× Concentrate is an RNase inhibitor.
A potent inhibitor of most nucleic acid binding enzymes, and thus useful as an RNase inhibitor. Especially useful when performing in situ hybridization. If it is added to all aqueous solutions used, it eliminates the need for special glassware washing and after-wash treatments. The 500x concentrate is economical; 2 ml treats 1,000 ml of solution. Not recommended in systems where other enzymatic activity is required.

Features and Benefits

  • The 500× concentrate is economical
  • Eliminates the need for special dishwashing and“after washing” cleaning and treatments.

Quantity

2mL treats 1,000ml of solution.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Legal Information

ProtectRNA is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential neurotoxicity of titanium implants: Prospective, in-vivo and in-vitro study
Shahar S, et al.
Biomaterials, 276, 121039-121039 (2021)
Exposing the Three-Dimensional Biogeography and Metabolic States of Pathogens in Cystic Fibrosis Sputum via Hydrogel Embedding, Clearing, and rRNA Labeling.
DePas WH
mBio, 7(5), e00796-e00716 (2016)
Intestinal epithelial cells express immunomodulatory ISG15 during active ulcerative colitis and Crohn?s disease
?stvik AE, et al.
Journal of Crohn's & Colitis, 14(7), 920-934 (2020)
RNA Sequencing of Intestinal Enterocytes Pre-and Post-Roux-en-Y Gastric Bypass Reveals Alteration in Gene Expression Related to Enterocyte Differentiation, Restitution, and Obesity with Regulation by Schlafen 12
Vomhof-DeKrey E E, et al.
Cells, 11(20), 3283-3283 (2022)
Recovery of high-quality RNA from laser capture microdissected human and rodent pancreas.
Butler AE
Journal of Histotechnology, 39(2), 59-65 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service