Skip to Content
Merck
All Photos(1)

Key Documents

R6901

Sigma-Aldrich

PCR 96 Well rack

assorted colors

Synonym(s):

microtube rack

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$146.00

$146.00


Estimated to ship onMay 23, 2025



Select a Size

Change View
5 EA
$146.00

About This Item

UNSPSC Code:
41122804
NACRES:
NB.15

$146.00


Estimated to ship onMay 23, 2025


material

polypropylene

feature

autoclavable
Holds 96 x 0.2 mL tubes

manufacturer/tradename

Heathrow Scientific HS23461A

technique(s)

PCR: suitable

size

127x99x30 mm

color

assorted colors

Looking for similar products? Visit Product Comparison Guide

General description

Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service