Skip to Content
Merck
All Photos(1)

Documents

R4529

Sigma-Aldrich

Anti-Rattin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Humanin-like in rat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

western blot: 1:500

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Rattin or RNR2 (RNA, ribosomal 45S cluster 2) is a subunit of the ribonucleoside diphosphate reductase. It is located on chromosome 14p12.

Immunogen

peptide corresponding to the rattin peptide (amino acids 20-35), which is not in the conserved region to humanin.

Biochem/physiol Actions

Rattin or RNR2 (RNA, ribosomal 45S cluster 2) helps in the synthesis of nucleotide precursors. It is responsible for the formation of the deoxyribonucleotides required for DNA synthesis. It is an important cofactor of HIV-1, that induces reverse transcription in MDMs (monocyte-derived macrophages).

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of yeast cells transformed with GFP expression constructs containing the RAD54 or RNR2 promoter as a test for the genotoxic potential of chemical substances
Afanassiev V, et al.
Mutation Research, 464(2), 297-308 (2000)
Identification of the DNA damage-responsive element of RNR2 and evidence that four distinct cellular factors bind it
Elledge SJ and Davis RW, et al.
Molecular and Cellular Biology, 9(12), 5373-5386 (1989)
Manipulation of human minichromosomes to carry greater than megabase-sized chromosome inserts
Kuroiwa Y, et al.
Nature Biotechnology, 18(10), 1086-1090 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service