Skip to Content
Merck
All Photos(1)

Key Documents

R3025

Sigma-Aldrich

Anti-Rho-GDI antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL

Synonym(s):

Anti-GDIA1, Anti-HEL-S-47e, Anti-NPHS8, Anti-RHOGDI, Anti-RHOGDI-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IP
WB
clone:
polyclonal
species reactivity:
mouse, bovine, human
citations:
9
technique(s):
immunoprecipitation (IP): 4 μg using 500 μg of Jurkat RIPA lysate
western blot: 0.5-1.0 μg/mL using RIPA lysates from human HL-60, HeLa, Hs294T, Jurkat and murine 3T3/NIH cells and mouse 3T3/A31

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

mouse, bovine, human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 4 μg using 500 μg of Jurkat RIPA lysate
western blot: 0.5-1.0 μg/mL using RIPA lysates from human HL-60, HeLa, Hs294T, Jurkat and murine 3T3/NIH cells and mouse 3T3/A31

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGDIA(396)
mouse ... Arhgdia(192662)

General description

Rho guanine nucleotide dissociation inhibitor I (RhoGDI), also known as Rho GDP dissociation inhibitor α (ARHGDIA) is encoded by the gene mapped to human chromosome 17q25.3-25-3. RhoGDI is the member of Rho/Rac protein family. The encoded protein consists of 204 amino acids and has a molecular mass of 23,421,000 Mr. In mammals, three RhoGDI isoforms exists: RhoGDIα, RhoGDIβ and RhoGDIγ. RhoGDIα is widely expressed. RhoGDIβ is mainly expressed in hematopoietic cells and RhoGDIγ is present in the brain, lung, kidney and testis.
Rho proteins are basically described as guanine nucleotide-binding proteins (GNBPs). RhoGDI (Rho guanine nucleotide dissociation inhibitor)/ARHGDIA (Rho guanine nucleotide dissociation inhibitor) has three subtypes namely- RhoGDIα (Rho GDP dissociation inhibitor α), RhoGDIβ (Rho GDP dissociation inhibitor β) and RhoGDIγ (Rho GDP dissociation inhibitor γ). RhoGDIα is expressed everywhere. It is located on human chromosome 17q25.3. Its protein has 204 amino acids and belongs to the Rho/Rac family.

Specificity

The antibody recognizes Rho-GDI and cross-reacts with D4-GDI.

Immunogen

GST fusion protein corresponding to full-length human Rho-GDI.

Application

Anti-Rho-GDI antibody produced in rabbit has been used in immunoblotting.
Anti-Rho-GDI has been used in western blotting.

Biochem/physiol Actions

Rho proteins control the actin and microtubule cytoskeleton. RhoGDIα is necessary for the spatial and temporal modulation of RhoGNBPs (guanine nucleotide-binding proteins). In HCC (hepatocellular carcinoma) patients, absence of ARHGDIA (Rho guanine nucleotide dissociation inhibitor) affects the prognosis and induces invasion and metastasis of HCC cells.

Physical form

Solution in 0.1 M Tris-glycine, pH 7.4, containing 0.15 M NaCl and 0.05% sodium azide.

Preparation Note

Purified using protein A

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and Mechanistic Insights into the Regulation of the Fundamental Rho Regulator RhoGDI? by Lysine Acetylation
Kuhlmann N, et al.
The Journal of Biological Chemistry, 291 (2016)
BNN27, a 17-Spiroepoxy Steroid Derivative, Interacts With and Activates p75 Neurotrophin Receptor, Rescuing Cerebellar Granule Neurons from Apoptosis
Pediaditakis I, et al.
Frontiers in Pharmacology, 7, 512-512 (2016)
Rac signaling in breast cancer: a tale of GEFs and GAPs
Wertheimer E, et al.
Cellular Signalling, 24(2), 353-362 (2012)
Kyu-Seon Oh et al.
Immunity, 47(2), 298-309 (2017-08-13)
Despite the widespread use of glucocorticoids (GCs), their anti-inflammatory effects are not understood mechanistically. Numerous investigations have examined the effects of glucocorticoid receptor (GR) activation prior to inflammatory challenges. However, clinical situations are emulated by a GC intervention initiated in
Mahamat Babagana et al.
Aging, 13(15), 19207-19229 (2021-08-15)
Age-associated low-grade sterile inflammation, commonly referred to as inflammaging, is a recognized hallmark of aging, which contributes to many age-related diseases. While tissue-resident macrophages are innate immune cells that secrete many types of inflammatory cytokines in response to various stimuli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service