Skip to Content
Merck
All Photos(1)

Documents

R0884

Sigma-Aldrich

T7 RNA Polymerase

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

RNA Polymerase T7, RNA Polymerase, T7 from E. coli HMS 174/pAR1219

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

grade

for molecular biology

form

buffered aqueous solution

mol wt

98.8 kDa

concentration

10,000-50,000 U/mL

UniProt accession no.

foreign activity

DNase and RNase, none detected

storage temp.

−20°C

Gene Information

bacteriophage T7 ... T7p07(1261050)

Looking for similar products? Visit Product Comparison Guide

General description

T7 RNA polymerase is highly specific for the bacteriophage T7 promoter and terminator sequences. It is extensively used to prepare RNA transcripts for stuctural and metabolic studies. The RNA transcripts can be converted to probes for sensitive hybridization detection studies. T7 polymerase and dideoxynucleotides can be used to directly sequence DNA.

Components

T7 RNA Polymerase is supplied as a solution of 100 mM NaCl, 50 mM Tris-HCl (pH 7.9), 0.1 mM EDTA, 0.1% Triton X-100, 1 mM DTT, and 50% (v/v) glycerol.

Unit Definition

One unit will catalyze the incorporation of 1 nmol of rNTP into acid-precipitable material in 60 min at 37°C.

Analysis Note

Activity assay: 40 mM Tris-HCl, pH 7.9, 6 mM MgCl2, 4 mM spermidine, 10 mM DTT, 0.5 μM each rNTP + 10 μCi α-32P-UTP, 3-10 units of enzyme, and 1 μg of a 350 bp template are incubated for 10 min at 37°C in a total volume of 100 μl. Typical results are ≥50% incorporation of labeled nucleotide into ≥90% full-length transcript.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V D Axelrod et al.
Biochemistry, 24(21), 5716-5723 (1985-10-08)
RNA synthesis by T7 RNA polymerase or SP6 RNA polymerase is 100-1000 times more sensitive to the presence of the 3'-deoxyribonucleoside 5'-triphosphate chain terminators than is RNA synthesis by Escherichia coli RNA polymerase or Q beta replicase. These ribonucleotide analogues
Pemphigus vulgaris is characterized by low IgG reactivities to specific self-antigens along with high IgG reactivity to desmoglein 3.
Fatta L
Immunology, 143(3), 374-380 (2014)
Albert Weixlbaumer et al.
Cell, 152(3), 431-441 (2013-02-05)
Transcriptional pausing by multisubunit RNA polymerases (RNAPs) is a key mechanism for regulating gene expression in both prokaryotes and eukaryotes and is a prerequisite for transcription termination. Pausing and termination states are thought to arise through a common, elemental pause
Tokiko Watanabe et al.
Journal of virology, 87(9), 5239-5254 (2013-03-02)
The 1918 pandemic influenza virus was the most devastating infectious agent in human history, causing fatal pneumonia and an estimated 20 to 50 million deaths worldwide. Previous studies indicated a prominent role of the hemagglutinin (HA) gene in efficient replication
Looking for a promoter in 3D.
Vladimir Svetlov et al.
Nature structural & molecular biology, 20(2), 141-142 (2013-02-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service