Skip to Content
Merck
All Photos(1)

Key Documents

PZ0360

Sigma-Aldrich

PF-4191834

≥98% (HPLC)

Synonym(s):

4-(3-(4-(1-Methyl-1H-pyrazol-5-yl) phenylthio) phenyl)-tetrahydro-2H-pyran-4-carboxamide, PF 4191834, Tetrahydro-4-[3-[[4-(1-methyl-1H-pyrazol-5-yl)phenyl]thio]phenyl]-2H-pyran-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$532.00

$132.00


Estimated to ship onApril 12, 2025



Select a Size

Change View
5 MG
$132.00
25 MG
$532.00

About This Item

Empirical Formula (Hill Notation):
C22H23N3O2S
CAS Number:
Molecular Weight:
393.50
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$132.00


Estimated to ship onApril 12, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

NC(C1(CCOCC1)C2=CC=CC(SC3=CC=C(C4=CC=NN4C)C=C3)=C2)=O

InChI

1S/C22H23N3O2S/c1-25-20(9-12-24-25)16-5-7-18(8-6-16)28-19-4-2-3-17(15-19)22(21(23)26)10-13-27-14-11-22/h2-9,12,15H,10-11,13-14H2,1H3,(H2,23,26)

InChI key

DVNQWYLVSNPCJZ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-4191834 a selective, non-redox, non-iron chelating competitive inhibitor of 5-lipoxygenase effective in inflammation and pain.
PF-4191834 is known to reduce the indications of arthritis.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaime L Masferrer et al.
The Journal of pharmacology and experimental therapeutics, 334(1), 294-301 (2010-04-10)
5-Lipoxygenase (LOX) is an important arachidonic acid-metabolizing enzyme producing leukotrienes and other proinflammatory lipid mediators with potent pathophysiological functions in asthma and other inflammatory diseases. 4-(3-(4-(1-Methyl-1H-pyrazol-5-yl)phenylthio)phenyl)-tetrahydro-2H-pyran-4-carboxamide (PF-4191834) is a novel, selective non-redox 5-lipoxygenase inhibitor effective in inflammation and pain. In
Han-Ching Lin et al.
PloS one, 9(9), e107890-e107890 (2014-09-18)
The lipoxygenase isoform of 5-lipoxygenase (5-LOX) is reported to be overexpressed in human rheumatoid arthritis synovial tissue and involved in the progress of inflammatory arthritis. However, the detailed mechanism of how 5-lipoxygenase regulates the inflammatory response in arthritis synovial tissue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service