Skip to Content
Merck
All Photos(1)

Key Documents

PZ0342

Sigma-Aldrich

PF-6274484

≥98% (HPLC)

Synonym(s):

N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00
25 MG
$465.00

$115.00


Estimated to ship onApril 27, 2025



Select a Size

Change View
5 MG
$115.00
25 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C18H14ClFN4O2
CAS Number:
Molecular Weight:
372.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Estimated to ship onApril 27, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

FC(C=C1)=C(Cl)C=C1NC2=NC=NC3=CC(OC)=C(NC(C=C)=O)C=C32

InChI

1S/C19H16ClFN4O/c1-4-11(2)24-17-8-13-16(9-18(17)26-3)22-10-23-19(13)25-12-5-6-15(21)14(20)7-12/h4-10,24H,1-2H2,3H3,(H,22,23,25)

InChI key

DTMQCGFVWWBONL-UHFFFAOYSA-N

Biochem/physiol Actions

PF-6274484 is an irreversible EGFR kinase inhibitor that covalently reacts with active-site cysteine residues in the ATP binding pocket. PF-6274484 inhibits autophosphorylation of both wild type and mutant EGFR in tumor cells with IC50 values of 5.8 nM and 6.6 nM, respectively.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service