Skip to Content
Merck
All Photos(1)

Key Documents

PZ0322

Sigma-Aldrich

WAY 170523

≥98% (HPLC)

Synonym(s):

N-[2-[4-[[[2-[(Hydroxyamino)carbonyl]-4,6-dimethylphenyl](phenylmethyl)amino]sulfonyl]phenoxy]ethyl]-2-benzofurancarboxamide, WAY170523

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$547.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$135.00
25 MG
$547.00

About This Item

Empirical Formula (Hill Notation):
C33H31N3O7S
CAS Number:
Molecular Weight:
613.68
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(N(CC1=CC=CC=C1)C2=C(C(NO)=O)C=C(C)C=C2C)(C3=CC=C(OCCNC(C4=CC5=C(C=CC=C5)O4)=O)C=C3)=O

InChI

1S/C33H31N3O7S/c1-22-18-23(2)31(28(19-22)32(37)35-39)36(21-24-8-4-3-5-9-24)44(40,41)27-14-12-26(13-15-27)42-17-16-34-33(38)30-20-25-10-6-7-11-29(25)43-30/h3-15,18-20,39H,16-17,21H2,1-2H3,(H,34,38)(H,35,37)

InChI key

FARMEEAGJWMFSZ-UHFFFAOYSA-N

Biochem/physiol Actions

WAY 170523 is a potent and selective inhibitor of matrix metalloprotease MMP-13 (Collagenase-3), expressed in the skeleton during embryonic development, and sometimes highly overexpressed in human carcinomas and in chondrocytes and synovial cells in rheumatoid arthritis and osteoarthritis. Mutations in the MMP-13 gene has been shown to cause metaphyseal anadysplasia (MANDP). WAY 170523 has an IC50 of 17 nM for MMP-13 and showed >5800-, 56-, and >500-fold selectivity against MMP-1, MMP-9, and TACE (TNFα converting enzyme), respectively.

Other Notes

This compound was developed by Pfizer for Cytoskeleton & Extracellular Matrix research. To learn more about Sigma′s partnership with Pfizer and view other authentic, high-quality Pfizer compounds, visit sigma.com/bsm-pfizer.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service