Skip to Content
Merck
All Photos(1)

Key Documents

PZ0272

Sigma-Aldrich

PF-06260933 dihydrochloride

≥98% (HPLC)

Synonym(s):

5-(4-Chloro-phenyl)-[3,3′]bipyridinyl-6,6′-diamine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$580.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$144.00
25 MG
$580.00

About This Item

Empirical Formula (Hill Notation):
C16H13ClN4 · 2HCl
CAS Number:
Molecular Weight:
369.68
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C(C2=CC=C(Cl)C=C2)C=C(C3=CN=C(N)C=C3)C=N1.[H]Cl

InChI

1S/C16H13ClN4.ClH/c17-13-4-1-10(2-5-13)14-7-12(9-21-16(14)19)11-3-6-15(18)20-8-11;/h1-9H,(H2,18,20)(H2,19,21);1H

InChI key

ZKIFOIROEWYGKV-UHFFFAOYSA-N

Biochem/physiol Actions

MAP4K4 inhibitor
PF-06260933 dihydrochloride is a MAP4K4 inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinpeng Li et al.
Cell death & disease, 8(8), e2965-e2965 (2017-08-05)
Endothelial-to-mesenchymal transition (EndMT) has been shown to contribute to organ fibrogenesis, and we have reported that the anti-EndMT effect of N-acetyl-seryl-aspartyl-lysyl-proline (AcSDKP) is associated with restoring expression of diabetes-suppressed fibroblast growth factor receptor (FGFR), the key anti-EndMT molecule. FGFR1 is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service