Skip to Content
Merck
All Photos(1)

Key Documents

PZ0245

Sigma-Aldrich

PF-05105679

≥98% (HPLC)

Synonym(s):

3-[[[(1R)-1-(4-Fluorophenyl)ethyl](3-quinolinylcarbonyl)amino]methyl]-benzoic acid, PF05105679, [(R)-3-[(1-(4-fluorophenyl)ethyl)(quinolin-3-ylcarbonyl)amino]methylbenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$148.00
25 MG
$580.00

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$148.00
25 MG
$580.00

About This Item

Empirical Formula (Hill Notation):
C26H21FN2O3
CAS Number:
Molecular Weight:
428.45
UNSPSC Code:
41116158
NACRES:
NA.77

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

PF-05105679 is an antagonist of the TRP (transient receptor potential) channel, also known as the cold and menthol receptor 1 (CMR1), with an IC50 value of 103 nM. PF-05105679 is in clinical trials for neuropathic pain. Participants taking PF-05105679 could hold their hands in a bucket of ice water for a long time before feeling pain, and pain-alleviating effects matched those of oxycodone for about one hour after administration.
PF-05105679 is an antagonist of the TRP (transient receptor potential) channel, also known as the cold and menthol receptor 1 (CMR1), with an IC50 value of 103 nM. PF-05105679 is in clinical trials for neuropathic pain. Participants taking PF-05105679 could hold their hands in a bucket of ice water for a long time before feeling pain, and pain-alleviating effects matched those of oxycodone for about one hour after administration.
Transient receptor potential cation channel subfamily M member 8 (TRPM8) antagonist, PF-05105679 [(R)-3-[(1-(4-fluorophenyl)ethyl)(quinolin-3-ylcarbonyl)amino]methylbenzoic acid] acts as a clinical tool to assess the function of TRPM8 in modulating the body temperature. It also helps to determine its role in the treatment of pain in humans.[1]
PF-05105679 is an antagonist of the TRP (transient receptor potential) channel, also known as the cold and menthol receptor 1 (CMR1).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a Selective TRPM8 Antagonist with Clinical Efficacy in Cold-Related Pain.
Andrews MD, et al.
ACS Medicinal Chemistry Letters, 6(4), 419-424 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service