Skip to Content
Merck
All Photos(1)

Key Documents

PZ0238

Sigma-Aldrich

CP-944629

≥98% (HPLC)

Synonym(s):

3-(1,1-Dimethylethyl)-6-[4-(2,4,5-trifluorophenyl)-5-oxazolyl]-1,2,4-triazolo[4,3-a]pyridine, 3-tert-Butyl-6-[4-(2,4,5-trifluorophenyl)-1,3-oxazol-5-yl][1,2,4]triazolo[4,3-a]pyridine, 3-tert-Butyl-6-[4-(2,4,5-trifluorophenyl)oxazol-5-yl]-[1,2,4]triazolo[4,3-a]pyridine, 5-(3-tert-Butyl-[1,2,4]triazolo[4,3-a]pyridin-6-yl)-4-(2,4,5-trifluorophenyl)oxazole, CP944629

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00

$122.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
5 MG
$122.00

About This Item

Empirical Formula (Hill Notation):
C19H15F3N4O
CAS Number:
Molecular Weight:
372.34
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00


Estimated to ship onApril 16, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C)(C)C1=NN=C2C=CC(C3=C(C4=C(F)C=C(F)C(F)=C4)N=CO3)=CN21

InChI

1S/C19H15F3N4O/c1-19(2,3)18-25-24-15-5-4-10(8-26(15)18)17-16(23-9-27-17)11-6-13(21)14(22)7-12(11)20/h4-9H,1-3H3

InChI key

VNZJNPIWZYMGAO-UHFFFAOYSA-N

Biochem/physiol Actions

CP-9446219 is a potent p38alpha (p38α) inhibitor (IC50 = 1.8 nM) with no inhibitory potency against a panel of 26 other knases (IC50 >10 μM), including p38gamma (p38γ) and p38delta (p38δ), MKK1, MAPK2/ERK2, JNK, MAPKAP-K1a, and MAPKAP-K2.
CP-9446219 is a potent p38alpha (p38α) inhibitor (IC50 = 1.8 nM) with no inhibitory potency against a panel of 26 other knases (IC50 >10 μM), including p38gamma (p38γ) and p38delta (p38δ), MKK1, MAPK2/ERK2, JNK, MAPKAP-K1a, and MAPKAP-K2. CP-944629 effectively inhibits LPS-stimulated cellular TNF-α production in vitro (IC50 = 63 and 8.3 nM, using human whole blood and isolated mononuclear cells, respectively) and in rats in vivo (ED50 = 0.1 mg/kg p.o.) with good pharmacokinetic profile and oral availability (F = 66%/monkey, 54%/rat).

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service