Skip to Content
Merck
All Photos(1)

Key Documents

PZ0209

Sigma-Aldrich

PF-06465469

≥97% (HPLC)

Synonym(s):

3-[4-Amino-1-[(3R)-1-(1-oxo-2-propen-1-yl)-3-piperidinyl]-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-N-[3-methyl-4-(1-methylethyl)phenyl]-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$161.00
25 MG
$623.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$161.00
25 MG
$623.00

About This Item

Empirical Formula (Hill Notation):
C30H33N7O2
CAS Number:
Molecular Weight:
523.63
UNSPSC Code:
12352200
NACRES:
NA.77

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1(C[C@@H](CCC1)[n]2nc(c5c2ncnc5N)c3cc(ccc3)C(=O)Nc4cc(c(cc4)C(C)C)C)C(=O)C=C

InChI

1S/C30H33N7O2/c1-5-25(38)36-13-7-10-23(16-36)37-29-26(28(31)32-17-33-29)27(35-37)20-8-6-9-21(15-20)30(39)34-22-11-12-24(18(2)3)19(4)14-22/h5-6,8-9,11-12,14-15,17-18,23H,1,7,10,13,16H2,2-4H3,(H,34,39)(H2,31,32,33)/t23-/m1/s1

InChI key

CGJVMKJGKFEHTL-HSZRJFAPSA-N

Biochem/physiol Actions

PF-06465469 is a potent, covalent inhibitor of the nonreceptor tyrosine kinase Itk (IL-2 inducible T-cell kinase), which is expressed primarily in immune cells, and is strongly upregulated following activation of T-cells. PF-06465469 inhibits Itk enzymatic activity with an IC50 of 2 nM. The compound blocks anti-CD3 induced phosphorylation of PLCg in Jurkat cells (IC50 = 31 nM) and IL-2 production in anti-CD3 stimulated human whole blood (IC50 = 48 nM).
PF-06465469 is a potent, covalent inhibitor of the nonreceptor tyrosine kinase Itk (IL-2 inducible T-cell kinase).

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Tec kinases respond to cellular stimulation by various receptors and are involved in signaling pathways.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service