PZ0177
OSU6162 hydrochloride
≥98% (HPLC)
Synonym(s):
PNU-96391; PNU-0096391; OSU-6162; (S)-(-)-(3-methanesulfonyl-phenyl)-1-propyl-piperidine hydrochloride; (S,S)-3-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine hydrochloride
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -5 to -7°, c = 1 in methanol
color
white to tan
solubility
H2O: ≥16 mg/mL
storage temp.
−20°C
SMILES string
Cl.CCCN1CCC[C@H](C1)c2cccc(c2)S(C)(=O)=O
InChI
1S/C15H23NO2S.ClH/c1-3-9-16-10-5-7-14(12-16)13-6-4-8-15(11-13)19(2,17)18;/h4,6,8,11,14H,3,5,7,9-10,12H2,1-2H3;1H/t14-;/m1./s1
InChI key
LEMGVHZVBREXAD-PFEQFJNWSA-N
Biochem/physiol Actions
Features and Benefits
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offer many products related to dopamine receptors for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service