Skip to Content
Merck
All Photos(1)

Key Documents

PZ0142

Sigma-Aldrich

PD-180970

≥98% (HPLC)

Synonym(s):

6-(2,6-Dichlorophenyl)-2-[(4-fluoro-3-methylphenyl)amino]-8-methyl-pyrido[2,3-d]pyrimidin-7(8H)-one, PF-1515965

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$195.00
25 MG
$769.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
5 MG
$195.00
25 MG
$769.00

About This Item

Empirical Formula (Hill Notation):
C21H15Cl2FN4O
CAS Number:
Molecular Weight:
429.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$195.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Assay

≥98% (HPLC)

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CN1C(=O)C(=Cc2cnc(Nc3ccc(F)c(C)c3)nc12)c4c(Cl)cccc4Cl

InChI

1S/C21H15Cl2FN4O/c1-11-8-13(6-7-17(11)24)26-21-25-10-12-9-14(20(29)28(2)19(12)27-21)18-15(22)4-3-5-16(18)23/h3-10H,1-2H3,(H,25,26,27)

InChI key

SLCFEJAMCRLYRG-UHFFFAOYSA-N

Biochem/physiol Actions

PD-180970 is a potent inhibitor of the p210 Bcr-Abl tyrosine kinase.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service