Skip to Content
Merck
All Photos(1)

Key Documents

PZ0042

Sigma-Aldrich

Sulopenem

≥97% (HPLC)

Synonym(s):

(5R,6S)-6-[(1R)-1-Hydroxyethyl]-7-oxo-3-[[(1R,3S)-tetrahydro-1-oxido-3-thienyl]thio]-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, CP-70429

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$537.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$537.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO5S3
CAS Number:
Molecular Weight:
349.45
UNSPSC Code:
51111800
NACRES:
NA.77

$132.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(O)C1=C(S[C@@H]2C[S@+]([O-])CC2)S[C@@]3([H])N1C([C@]3([H])[C@H](O)C)=O

InChI

1S/C12H15NO5S3/c1-5(14)7-9(15)13-8(11(16)17)12(20-10(7)13)19-6-2-3-21(18)4-6/h5-7,10,14H,2-4H2,1H3,(H,16,17)/t5-,6?,7+,10-,21?/m1/s1

InChI key

FLSUCZWOEMTFAQ-JNCILSLHSA-N

Biochem/physiol Actions

Orally available penem antibiotic to treat Gram-negative multi-drug resistant infections
Sulopenem is an orally available penem antibiotic designed to treat Gram-negative multi-drug resistant infections. It can be administered as an intravenous preparation, and it is also orally available.
Sulopenem, formerly CP-70,429, is stable to renal dehydropeptidase I and hydrolytic activity of AmpC β-lactamases and extended-spectrum β-lactamases (ESBLs) that convey resistance to third-generation cephalosporins. This action of sulopenem helps to act against urgent drug-resistant antimicrobial threats, including ESBL-producing Enterobacteriaceae.[1] Sulopenem is effective than ciprofloxacin for treating urogenital upper urinary tract infection (uUTI) in adult women due to quinolone non-susceptible pathogens.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James A Karlowsky et al.
Antimicrobial agents and chemotherapy, 63(1) (2018-11-07)
The in vitro activity of sulopenem was assessed against a collection from 2014 to 2016 of 539 urinary isolates of Escherichia coli from Canadian patients by using CLSI-defined broth microdilution methodology. A concentration of sulopenem 0.03 µg/ml inhibited both 50% (MIC50)
Simi S Paknikar et al.
North American journal of medical sciences, 4(11), 537-547 (2012-11-28)
In order to deal with the rising problem of antibiotic resistance, newer antibacterials are being discovered and added to existing pool. Since the year 2000, however, only four new classes of antibacterials have been discovered. These include the oxazolidinones, glycolipopeptides
LB-1. Efficacy and Safety of Oral Sulopenem Etzadroxil/Probenecid Versus Oral Ciprofloxacin in the Treatment of Uncomplicated Urinary Tract Infections (uUTI) in Adult Women: Results from the SURE-1 Trial
Open forum infectious diseases, 7(Suppl 1) , S844-S844 (2020)
Klaudia Kosowska-Shick et al.
Antimicrobial agents and chemotherapy, 53(6), 2239-2247 (2009-03-25)
For 297 penicillin-susceptible, -intermediate, and -resistant pneumococcal strains, the sulopenem MIC(50)s were 0.008, 0.06, and 0.25, respectively, and the sulopenem MIC(90)s were 0.016, 0.25, and 0.5 microg/ml, respectively. The MIC(50)s of amoxicillin for the corresponding strains were 0.03, 0.25, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service