PZ0022
BLI-489 hydrate
≥98% (HPLC)
Synonym(s):
6-[(5,6-Dihydro-8H-imidazo[2,1-c][1,4]oxazin-2-yl)methylene]-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid sodium salt hydrate, WAY-185489
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
yellow
solubility
H2O: >20 mg/mL
storage temp.
room temp
SMILES string
O.[Na+].[O-]C(=O)C1=CS[C@H]2N1C(=O)\C2=C\c3cnc4COCCn34
InChI
1S/C13H11N3O4S.Na.H2O/c17-11-8(12-16(11)9(6-21-12)13(18)19)3-7-4-14-10-5-20-2-1-15(7)10;;/h3-4,6,12H,1-2,5H2,(H,18,19);;1H2/q;+1;/p-1/b8-3-;;/t12-;;/m1../s1
InChI key
UAZCFKAYAYJPDM-UVEQJXRBSA-M
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service