Skip to Content
Merck
All Photos(1)

Key Documents

PURN12050

Sigma-Aldrich

Pur-A-Lyzer Mini Dialysis Kit

Mini 12000, capacity 10-250 μL, MWCO 12-14 kDa

Synonym(s):

Dialyzers, High through put dialysis tubing, puralyzer

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$446.00

$446.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 KIT
$446.00

About This Item

UNSPSC Code:
41123100
NACRES:
NB.22

$446.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

regenerated cellulose membrane

packaging

pkg of 50 Mini dialysis tubes

capacity

10-250 μL

tube size × diam.

3.5 cm × 0.9 cm

pore size

12-14 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Pur-A-Lyzer Dialysis Kits are an innovative dialysis tubing that can be conveniently loaded and unloaded using any standard pipette tip while still allowing for 98% sample recovery yield. The revolutionary design of the Pur-A-Lyzer Tube makes it the only dialysis tool which enables true high throughput dialysis, accommodating a large number of samples in a single buffer tank.

The Pur-A-Lyzer Dialysis Tube allows rapid, high performance extraction of macromolecules without contamination. It is ideal for purification of proteins, automated protein sequencing, peptide mapping and amino acid analysis.

Application

Dialysis or buffer exchange of volumes between 10 mL and 20 mL
Preparation of protein samples for MALDI-MS.
Sample concentration.
Large-scale protein dialysis such as antibodies and recombinant protein purification.
Peptide dialysis for molecules as small as 10 amino acids.
Virus-particle purification.
Removal of contaminating micro molecules.
Purification of tissue culture extracts.
Removal of salts, surfactants, solvents and detergents.
Complex formation studies (protein-protein, protein-DNA and protein-RNA)
pH and buffer adjustment of sample solution, protein extract or cell extract.
High through- put dialysis, a single beaker can be used for several Pur-A-Lyzer tubes on one floating pad.

For your convenience the Pur-A-Lyzer kits come in four sizes:
Mini: 10 - 250 μL
Midi: 50 - 800 μL
Maxi:0.1 - 3 mL
Mega: 3 - 20 mL

Legal Information

Pur-A-Lyzer is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service