Skip to Content
Merck
All Photos(3)

Documents

PRS3985

Sigma-Aldrich

Anti-Nicastrin (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-APH-2, Anti-Anterior pharynx defective 2, Anti-Nicastrin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCSTN(23385)

General description

Nicastrin (NCSTN) is a transmembrane glycoprotein, that is located on chromosome 1q23.

Immunogen

a 18 amino acid peptide from near the center of human Nicastrin.

Application

Anti-Nicastrin has been used in western blotting.

Biochem/physiol Actions

Nicastrin (NCSTN) helps in the assembly of γ-secretase complex. It is associated with Alzheimer′s disease.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3985.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of the novel activity-driven interaction between synaptotagmin 1 and presenilin 1 links calcium, synapse, and amyloid beta
Kuzuya A, et al.
BMC Biology, 14:25 (2016)
The gene encoding nicastrin, a major gamma-secretase component, modifies risk for familial early-onset Alzheimer disease in a Dutch population-based sample
Dermaut B, et al.
American Journal of Human Genetics, 70(6), 1568-1574 (2002)
Akira Kuzuya et al.
BMC biology, 14, 25-25 (2016-04-03)
Synaptic loss strongly correlates with memory deterioration. Local accumulation of amyloid β (Aβ) peptide, and neurotoxic Aβ42 in particular, due to abnormal neuronal activity may underlie synaptic dysfunction, neurodegeneration, and memory impairments. To gain an insight into molecular events underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service