Skip to Content
Merck
All Photos(3)

Key Documents

PRS3135

Sigma-Aldrich

Anti-TLR2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MAL, Anti-TIR domain containing adaptor protein, Anti-TIRAP, Anti-Toll-like receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLR2(7097)

Immunogen

a peptide corresponding to 14 amino acids near the amino-terminus of human TLR2.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3135.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianbing Ding et al.
Reproductive sciences (Thousand Oaks, Calif.), 25(5), 662-673 (2017-11-21)
Infectious agents are a significant risk factor for preterm birth (PTB); however, the simple presence of bacteria is not sufficient to induce PTB in most women. Human and animal data suggest that environmental toxicant exposures may act in concert with
Donghui Cao et al.
Carcinogenesis, 40(2), 234-245 (2018-10-27)
The natural phenolic substance, 18β-glycyrrhetinic acid (GRA), has shown enormous potential in the chemoprevention of cancers with rich resources and biological safety, but the GRA-regulated genetic and epigenetic profiles are unclear. Deregulated mitochondrial cellular energetics supporting higher adenosine triphosphate provisions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service