Skip to Content
Merck
All Photos(3)

Key Documents

PRS2221

Sigma-Aldrich

Anti-BAFF antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BLyS, Anti-Tall, Anti-Thank, Anti-Tumor necrosis factor (ligand) superfamily, member 13b

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 31 kDa

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

B-lymphocyte activating factor (BAFF/TNFSF13B) is a cytokine that belongs to the TNF (tumor necrosis factor) superfamily. BAFF is located on human chromosome 13q33.

Immunogen

a peptide corresponding to amino acids near the carboxy-terminus of human BAFF.

Biochem/physiol Actions

B cell activating factor (BAFF) participates in B cell and T cell mediated immunity. It plays a key role in the survival and differentiation of B cells. Mutations in BAFF regulates the levels of antibody. BAFF also participates in enhancing the responses of IL-6 (interlukin-6) and TNF-α (tumor necrosis factor alpha), that helps hepatocytes in the production of acute-phase protein.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2221.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of genetic variation in B-cell activating factor with chronic hepatitis B virus infection.
Han Q, et al.
Immunology Letters, 188, 53-58 (2017)
Sung-Sik Yoon et al.
International journal of molecular sciences, 22(13) (2021-07-21)
Fibroblast-like synoviocytes (FLSs) play a key role in the pathogenesis of rheumatoid arthritis (RA) by producing inflammatory cytokines and interacting with various immune cells, which contribute to cartilage destruction. RA-FLSs activated by tumor necrosis factor alpha (TNF-α), exacerbate joint damage
B cell-activating factor regulates the survival of B lymphocytes infected with human cytomegalovirus.
Xu H, et al.
Immunology Letters, 187, 1-6 (2017)
Increased levels of BAFF in patients with systemic lupus erythematosus are associated with acute-phase reactants, independent of BAFF genetics: a case?control study.
Eilertsen G?, et al.
Rheumatology (Basel), 50(12), 2197-2205 (2011)
Polymorphism in the 5′ regulatory region of the B-lymphocyte activating factor gene is associated with the Ro/La autoantibody response and serum BAFF levels in primary Sjogren's syndrome.
Nossent JC, et al.
Rheumatology (Basel), 47(9), 1311-1316 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service