Skip to Content
Merck
All Photos(3)

Documents

PLA0272

Sigma-Aldrich

Rabbit anti-USP8 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

HumORF8, KIAA0055, SPG59, UBPY, Ubiquitin thioesterase 8, deubiquitinating enzyme 8, hUBPy, ubiquitin isopeptidase Y, ubiquitin specific protease 8, ubiquitin thiolesterase 8, ubiquitin-specific-processing protease 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:1,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_005145.3

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... USP8(9101)

Immunogen

The epitope recognized by PLA0272 maps to a region between residue 400 and 450 of human Ubiquitin Specific Peptidase 8 using the numbering given in entry NP_005145.3 (GeneID 9101).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

USP8 (ubiquitin-specific processing protease 8) is also known as UBPY (ubiquitin isopeptidase Y). USP8 is a hydrolase that can remove conjugated ubiquitin from proteins and therefore plays an important regulatory role at the level of protein turnover by preventing degradation [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/ P40818].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service