Skip to Content
Merck
All Photos(1)

Key Documents

PLA0271

Sigma-Aldrich

Rabbit anti-GTF2B/TFIIB Antibody, Affinity Purified

Synonym(s):

RNA polymerase II transcription factor IIB, S300-II, TF2B, TFIIB, general transcription factor TFIIB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2- 5 μg/mg

accession no.

NP_001505.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Immunogen

The epitope recognized by PLA0271 maps to a region between residue 100 and 150 of human General Transcription Factor IIB using the numbering given in entry NP_001505.1 (GeneID 2959).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

GTF2B/TFIIB (General transcription factor IIB) is one of the ubiquitous factors required for transcription initiation by RNA polymerase II. GTF2B/TFIIB localizes to the nucleus where it forms a complex (the DAB complex) with transcription factors IID and IIA. Transcription factor IIB serves as a bridge between IID, the factor which initially recognizes the promoter sequence, and RNA polymerase II [taken from NCBI Entrez Gene (Gene ID: 2959)].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service