Skip to Content
Merck
All Photos(2)

Documents

PLA0130

Sigma-Aldrich

Rabbit anti-ATRIP Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATM and Rad3-related-interacting protein, FLJ12343, MGC20625, MGC21482, MGC26740

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_569055.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... ATRIP(84126)

Immunogen

The epitope recognized by PLA0130 maps to a region between residue 100 to 150 of human ATM and Rad3-related-interacting protein using the numbering given in entry NP_569055.1 (GeneID 84126).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

ATRIP (ATR-interacting protein) forms a complex with the serine-threonine kinase ATR (ataxia-telangiectasia mutated rad-3 related) to function in the DNA damage response. Through its interaction with replication protein A (RPA), ATRIP aids the recruitment of ATR to regions of DNA damage. ATR-ATRIP interacts with and is regulated by Rad17 and the 9-1-1 complexes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service