Skip to Content
Merck
All Photos(1)

Documents

PLA0010

Sigma-Aldrich

Goat anti-RENT1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATP-dependent helicase RENT1, HUPF1, KIAA0221, NORF1, RENT1, UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog, UPF1 regulator of nonsense transcripts homolog (yeast), delta helicase, nonsense mRNA reducing factor 1, nonsense mediated mRNA decay factor, nonsense mediated mRNA decay factor (C. elegans), pNORF1, regulator of nonsense transcripts 1, smg-2, smg-2 homolog, up-frameshift mutation 1 homolog, up-frameshift suppressor 1 homolog, yeast Upf1p homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): 2-10 μg/mL
western blot: 1:2,000-1:10000

accession no.

NP_002902.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... UPF1(5976)

Immunogen

The epitope recognized by PLA0010 maps to a region between residue 1075 and the C-terminus (residue 1118) of human Regulator of Nonsense Transcripts 1 (up-frameshift mutation 1 homolog) using the numbering given in entry NP_002902.2 (GeneID 5976).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

RENT1, also known as UPF1, is a key regulator of the nonsense-mediated mRNA decay (NMD) pathway. The NMD pathway eliminates mRNAs that prematurely terminate during translation and may serve to protect the cells from dominant-negative effects of truncated proteins, or as a means to regulate protein expression. RENT1 is an ATP-dependent 5′- 3′ RNA helicase and ATPase that localizes to the cytoplasm and is part of a post-splicing multiprotein complex as well as an mRNA decay complex.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service