Skip to Content
Merck

PCG3003

TruPAGE Tris-MOPS SDS Express Running Buffer

20 ×

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.22
UNSPSC Code:
12352201

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

concentration

20 ×

compatibility

for use with Sigma-Aldrich TruPAGE Precast Gels

storage temp.

room temp

Application

TruPAGE Tris-MOPS SDS Express Running Buffer has been used to separate proteins through sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE).[1]

Features and Benefits

TruPAGE Tris-MOPS SDS Express Running Buffer is specially formulated to be used with TruPAGE Precast Gels. This running buffer will provide increased resolution for small MW proteins, and will noticeably shorten the run time. This running buffer needs to be supplemented with a 1X working concentration of TruPAGE Antioxidant (PCG3007) to prevent protein reoxidation during electrophoresis when running reduced protein samples. Please refer to the gel migration chart to select the appropriate precast gel and running buffer combination for your experimental needs.

Legal Information

TruPAGE is a trademark of Sigma-Aldrich Co. LLC

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of phosphorylated Akt and MAPK in cell culture assays.
Molgaard S, et al.
MethodsX, 3, 386-398 (2016)
Simon Molgaard et al.
MethodsX, 3, 386-398 (2016-06-09)
This article describes an immunocytochemistry (ICC) method for staining against phosphorylated forms of the kinases Akt (pAkt) and MAPK (pMAPK). Phosphorylation is induced upon their activation by a number stimuli including insulin and brain-derived neurotrophic factor (BDNF), and is prerequisite
John R Christin et al.
Cell reports, 31(10), 107742-107742 (2020-06-11)
Lineage plasticity is important for the development of basal-like breast cancer (BLBC), an aggressive cancer subtype. While BLBC is likely to originate from luminal progenitor cells, it acquires substantial basal cell features and contains a heterogenous collection of cells exhibiting

Global Trade Item Number

SKUGTIN
PCG3003-500ML04061838262141
PCG3003-4L04061834406426

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service