P9192
Pneumadin Rat
≥97% (HPLC)
Synonym(s):
Tyr-Gly-Glu-Pro-Lys-Leu-Asp-Ala-Gly-Val-NH2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C47H74N12O15
CAS Number:
Molecular Weight:
1047.16
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CC(C)C[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCC(O)=O)NC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](C(C)C)C(N)=O
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Markowska et al.
Histology and histopathology, 12(3), 677-682 (1997-07-01)
Pneumadin (PNM) is a biologically active decapeptide, which has previously been found to enhanced rat adrenal growth; the mechanism is indirect and probably involves the stimulation of both arginine-vasopressin (AVP) and ACTH release. The effects of 2- and 6-day PNM
V K Batra et al.
Biochemical pharmacology, 58(1), 177-182 (1999-07-14)
The direct vascular effect of pneumadin (PN) was determined by studying the changes in intracellular free calcium ([Ca2+]i) levels in cultured rat aortic smooth muscle cells maintained between the second and fifth passages. PN evoked a rapid, concentration-dependent, biphasic increase
J D Watson et al.
Regulatory peptides, 57(2), 105-114 (1995-05-30)
Pneumadin is an antidiuretic decapeptide, recently isolated from rat and human lung. Bolus intravenous injection of 5 nmol of pneumadin into water-loaded rats caused a rapid and significant antidiuresis and a reduction in Na+ and Cl- excretion. Pneumadin administration did
J Kosowicz et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 36(2), 78-81 (2004-03-06)
Pneumadin (PNM) is a decapeptide originally isolated from mammalian lungs, and exerts a potent antidiuretic action by stimulating arginine-vasopressin release. We have recently developed a sensitive and specific radioimmunoassay (RIA) for rat PNM and detected high concentrations of PNM--not only
A Markowska et al.
Histology and histopathology, 11(3), 583-585 (1996-07-01)
Pneumadin (PNM) is a biologically active decapeptide, originally isolated from mammalian lungs, that has been previously found to acutely stimulate pituitary-adrenocortical axis in rats. The effects of 2-day PNM administration on the atrophic adrenal cortices of rats treated for 8
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service