Skip to Content
Merck
All Photos(1)

Key Documents

P8913

Sigma-Aldrich

Phospholipase A2 from bovine pancreas

lyophilized powder, ≥20 units/mg protein

Synonym(s):

Lecithinase A, PLA2, Phosphatidylcholine 2-acylhydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$207.00
10 MG
$356.00
50 MG
$1,190.00

$207.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$207.00
10 MG
$356.00
50 MG
$1,190.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

$207.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

form

lyophilized powder

Quality Level

specific activity

≥20 units/mg protein

composition

Protein, 5-20% biuret

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Phospholipase A2 from bovine pancreas is a calcium-dependent lipolytic enzyme.[1]

Application

Phospholipase A2 from bovine pancreas has been used to determine rat renal proximal tubular segments (PTS) viability during oxygenation and hypoxia-reoxygenation[2] and to study the impact of phospholipase A2 on tubular cell triglyceride levels.[3][4]

Biochem/physiol Actions

Hydrolyzes the β-ester bond of zwitterionic glycerophospholipids. Preferred substrates are phosphatidylcholine, phosphatidylethanolamine, and their plasmalogen analogues. Phosphatidylinositol and phosphatidylserine are also hydrolyzed. Aggressively attacks phospholipids in membranes of intact cells.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of soybean L-α-phosphatidylcholine to L-α-lysophosphatidylcholine and a fatty acid per min at pH 8.0 and 37 °C.

Physical form

Contains Tris buffer salts.

Analysis Note

Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Triglyceride accumulation in injured renal tubular cells: alterations in both synthetic and catabolic pathways.
Johnson AC
Kidney International, 67(6), 2196-2209 (2005)
Renal tubular triglyercide accumulation following endotoxic, toxic, and ischemic injury.
Zager RA
Kidney International, 67(1), 111-121 (2005)
Phospholipase A2 activity can protect renal tubules from oxygen deprivation injury.
Zager RA
Proceedings of the National Academy of Sciences of the USA, 90(17), 8297-8301 (1993)
Ming Chen et al.
Scientific reports, 7(1), 15931-15931 (2017-11-23)
Hepatitis C virus (HCV), dengue virus (DENV) and Japanese encephalitis virus (JEV) belong to the family Flaviviridae. Their viral particles have the envelope composed of viral proteins and a lipid bilayer acquired from budding through the endoplasmic reticulum (ER). The
K Sekar et al.
Acta crystallographica. Section D, Biological crystallography, 62(Pt 4), 392-397 (2006-03-23)
The lipolytic enzyme phospholipase A2 plays a crucial role in lipid metabolism and catalyzes hydrolysis of the fatty-acid ester bond at the sn-2 position of phospholipids. Here, the crystal structure (1.7 A resolution) of the triple mutant (K53,56,121M) of bovine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service