Skip to Content
Merck
All Photos(1)

Key Documents

P8828

Sigma-Aldrich

Pergolide mesylate salt

≥98%, solid

Synonym(s):

8β-[(Methylthio)methyl]-6-propylergoline methanesulfonate salt, Pergolide methanesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26N2S · CH4O3S
CAS Number:
Molecular Weight:
410.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

solid

optical activity

[α]20/D −35°, c = 0.2 in pyridine(lit.)

color

white

solubility

ethanol: 2 mg/mL
DMSO: 5.2 mg/mL
H2O: insoluble

originator

Valeant

storage temp.

−20°C

SMILES string

CS(O)(=O)=O.[H][C@@]12Cc3c[nH]c4cccc(c34)[C@@]1([H])C[C@@H](CSC)CN2CCC

InChI

1S/C19H26N2S.CH4O3S/c1-3-7-21-11-13(12-22-2)8-16-15-5-4-6-17-19(15)14(10-20-17)9-18(16)21;1-5(2,3)4/h4-6,10,13,16,18,20H,3,7-9,11-12H2,1-2H3;1H3,(H,2,3,4)/t13-,16-,18-;/m1./s1

InChI key

UWCVGPLTGZWHGS-ZORIOUSZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopaminergic agonist; suppresses pituitary secretion of prolactin; anti-Parkinsonian agent.

Features and Benefits

This compound was developed by Valeant. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dianne McFarlane
The Veterinary clinics of North America. Equine practice, 27(1), 93-113 (2011-03-12)
Equine pituitary pars intermedia dysfunction (PPID), also known as equine Cushing's syndrome, is a widely recognized disease of aged horses. Over the past two decades, the aged horse population has expanded significantly and in addition, client awareness of PPID has
Kamal S Bhatia et al.
Behavioural brain research, 223(1), 227-232 (2011-05-17)
Psychostimulant abuse represents a psychiatric disorder and societal concern that has been largely unamenable to therapeutic interventions. We have previously demonstrated that the 5-HT₃ antagonist ondansetron or non-selective 5-HT(₂A/₂C) antagonist ketanserin administered 3.5 h following daily pergolide, a non-selective DA
Hans C Lou et al.
Journal of vision, 11(2), doi:10-doi:10 (2011-02-25)
Liberal acceptance, overconfidence, and increased activity of the neurotransmitter dopamine have been proposed to account for abnormal sensory experiences, for instance, hallucinations in schizophrenia. In normal subjects, increased sensory experience in Yoga Nidra meditation is linked to striatal dopamine release.
P Barone et al.
Neurology, 53(3), 573-579 (1999-08-17)
To determine whether pergolide monotherapy provides symptomatic relief in early PD. Early treatment with dopamine agonists may reduce the risk of motor fluctuations, which are most likely linked to levodopa therapy. Pergolide, a D1-D2 dopamine agonist, has been studied as
Nathan Silver et al.
Sleep medicine, 12(5), 440-444 (2011-01-18)
Restless legs syndrome (RLS) is a chronic disease, which is managed with palliative medications that are likely to be required for a patient's lifetime. It is, therefore, important to know the long-term consequences of these treatments. Currently, the most commonly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service