Skip to Content
Merck
All Photos(1)

Key Documents

P8374

Sigma-Aldrich

Pyoverdines−Fe complex

from Pseudomonas fluorescens, >90% (HPLC)

Synonym(s):

Pyoverdines, Fe-Pv

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$378.00

$378.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$378.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$378.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

biological source

Pseudomonas fluorescens

Quality Level

Assay

>90% (HPLC)

form

powder

solubility

H2O: ~10 mg/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pyoverdines-Fe complex has been used in cyclic voltammetry to determine potential detection windows and peak potentials.[1][2]

Biochem/physiol Actions

Pyoverdines, also called pseuobactins and pyoverdins, are fluorescent siderophores that have high affinity for iron (1032 M-1), and are synthesized by fluorescent pseudomonads under iron-deficient growth conditions. Pyoverdines were shown to prevent iron toxicity produced by iron overload in hepatocyte cultures and effectively scavenges the hydroxyl and peroxyl radicals. Pyoverdines are effective in acquiring iron from transferrin and lactoferrin. Pyoverdines are also involved in the suppression of pythium-induced damping-off of tomato and promotion of growth in some higher plants.
Pyoverdines, also called pseuobactins and pyoverdins, are fluorescent siderophores that have high affinity for iron, and are synthesized by fluorescent pseudomonads under iron-deficient growth conditions.
The complex ferri-pyoverdine acts as a signaling molecule inducing the production of secreted virulence factors in Pseudomonas aeruginosa.[3] Ferri-pyoverdines promote the growth of both P. fluorescens and P. aeruginosa in minimal medium.

Physical form

Supplied as a mixture containing mainly the succinic acid (MW=1161), 2-hydroxy glutaramide (MW=1190), and succinamide (MW=1160) forms of pyoverdines, bound to iron.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast Selective Detection of Pyocyanin Using Cyclic Voltammetry
Sensors (2016)
Paul A Beare et al.
Molecular microbiology, 47(1), 195-207 (2002-12-21)
Under iron-limiting conditions, Pseudomonas aeruginosa produces a siderophore called pyoverdine. Pyoverdine is secreted into the extracellular environment where it chelates iron, and the resulting ferri-pyoverdine complexes are transported back into the bacteria by a cell surface receptor protein FpvA. Pyoverdine
Electrochemical sensing of biomarker for diagnostics of bacteria-specific infections
Fatima A
Nanomedicine (London, England) (2016)
Islem Gandouzi et al.
Materials (Basel, Switzerland), 12(7) (2019-04-14)
Pyoverdine is a fluorescent siderophore produced by Pseudomonas aeruginosa that can be considered as a detectable marker in nosocomial infections. The presence of pyoverdine in water can be directly linked to the presence of the P. aeruginosa, thus being a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service