Skip to Content
Merck
All Photos(2)

Key Documents

P8122

Sigma-Aldrich

Anti-Potassium Channel KIR3.2 (GIRK2) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BIR1, Anti-GIRK-2, Anti-GIRK2, Anti-KATP-2, Anti-KATP2, Anti-KCNJ7, Anti-KIR3.2, Anti-KPLBS, Anti-hiGIRK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ6(3763)
mouse ... Kcnj6(16522)
rat ... Kcnj6(25743)

Related Categories

General description

G protein-activated inward rectifier potassium channel 2 (GIRK2) is localized on the plasma membrane. It is expressed majorly in the brain and to some extent in the pancreas. The GIRK2 gene is mapped to human chromosome 21q22.13. Structurally, GIRK2 exists as a tetramer and comprises a transmembrane domain and a cytoplasmic domain.

Specificity

Anti-Potassium Channel KIR3.2 (GIRK2) recognizesKIR3.2 (GIRK2).

Immunogen

GST fusion protein with sequence corresponding to residues 374-414 of mouse KIR3.2 (GIRK2). Sequence homology is 40/41, 39/41, and 37/41 residues identical in rat, golden hamster, and human, respectively.

Application

Anti-Potassium Channel KIR3.2 (GIRK2) antibody produced in rabbit has been used in:
  • immunoblotting[1]
  • immunofluorescence[2][3]
  • immunocytochemistry

Biochem/physiol Actions

G protein-activated inward rectifier potassium channel 2,(GIRK2) plays a key role in γ-aminobutyric acid-ergic (GABAergic), dopaminergic, cholinergic, and glutamatergic synapses. It also regulates neuronal excitability. GIRK2 through G protein-coupled receptor stimulation modulates neuronal circuit activity and heart rate in cardiac cells. Deletion of the GIRK2 gene in mice may lead to an anxiety-like phenotype and a depression-resistant behavior.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kumar et al.
Anaesthesia, 74(11), 1456-1470 (2019-08-28)
Advances in the field of pharmacogenomics have resulted in the discovery of some important single-nucleotide polymorphisms which are found to be associated with opioid dose variability. This, to a large extent, explains genetic variability in the analgesic dose of opioids.
Nerea Llamosas et al.
The international journal of neuropsychopharmacology, 18(11), pyv051-pyv051 (2015-05-10)
Targeting dorsal raphe 5-HT1A receptors, which are coupled to G-protein inwardly rectifying potassium (GIRK) channels, has revealed their contribution not only to behavioral and functional aspects of depression but also to the clinical response to its treatment. Although GIRK channels
Matthew R Whorton et al.
Cell, 147(1), 199-208 (2011-10-04)
G protein-gated K(+) channels (Kir3.1-Kir3.4) control electrical excitability in many different cells. Among their functions relevant to human physiology and disease, they regulate the heart rate and govern a wide range of neuronal activities. Here, we present the first crystal
Chella Kamarajan et al.
International journal of psychophysiology : official journal of the International Organization of Psychophysiology, 115, 13-23 (2016-12-21)
Event related oscillations (EROs) are heritable measures of neurocognitive function that have served as useful phenotype in genetic research. A recent family genome-wide association study (GWAS) by the Collaborative Study on the Genetics of Alcoholism (COGA) found that theta EROs
Carlo Corona et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(27), 6102-6113 (2018-06-08)
Activating Transcription Factor 4 (ATF4) has been postulated as a key regulator of learning and memory. We previously reported that specific hippocampal ATF4 downregulation causes deficits in synaptic plasticity and memory and reduction of glutamatergic functionality. Here we extend our

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service