Skip to Content
Merck
All Photos(1)

Documents

P8049

Sigma-Aldrich

Pipette Pump II

blue

Synonym(s):

pipette filler, thumb wheel pipette pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

plastic
silicone

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37911-1002

volume

2 mL

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

For safe and accurate pipetting with one-handed operation. Rotation of the thumb wheel provides precise control of filling or dispensing. Rapid emptying is possible by pressing the plunger or the quick release bar (not available on the 0.2 mL pump). Color coded for easy identification.

Compatibility

for pipettes up to 2 mL

Legal Information

Pipette Pump is a trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service