Skip to Content
Merck
All Photos(1)

Key Documents

P7903

Sigma-Aldrich

Pipemidic acid

Synonym(s):

8-Ethyl-5,8-dihydro-5-oxo-2-(1-piperazinyl)pyrido(2,3-d)pyrimidine-6-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N5O3
CAS Number:
Molecular Weight:
303.32
EC Number:
MDL number:
UNSPSC Code:
51282938
PubChem Substance ID:
NACRES:
NA.85

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
cell wall synthesis | interferes
enzyme | inhibits

SMILES string

CCN1C=C(C(O)=O)C(=O)c2cnc(nc12)N3CCNCC3

InChI

1S/C14H17N5O3/c1-2-18-8-10(13(21)22)11(20)9-7-16-14(17-12(9)18)19-5-3-15-4-6-19/h7-8,15H,2-6H2,1H3,(H,21,22)

InChI key

JOHZPMXAZQZXHR-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

General description

Chemical structure: quinolone

Application

Pipemidic acid is used to study DNA damage and mutagenic activity. It has been used to induce articular lesions in the neonatal mouse and is used to study human lymphocyte blastogenesis and γ-interferon production .

Biochem/physiol Actions

Pipemidic acid modulates (inhibits) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent) DNA supercoiling, and chromosome fragmentation. Pipemidic acid has been shown to induce inhibition of lymphocyte DNA synthesis.

Analysis Note

Very slightly soluble in water. It dissolves in diluted solutions of acids and of alkali hydroxides.

Other Notes

10g,100g
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao-Dong Liang et al.
Analytical and bioanalytical chemistry, 380(7-8), 918-923 (2004-11-30)
A flow-injection chemiluminescence (CL) method for the determination of pipemidic acid is described. It is based on energy transfer from excited state peroxynitrous acid to pipemidic acid, in which the excited state peroxynitrous acid is synthesized on-line by the mixing
Margherita Lavorgna et al.
International journal of molecular sciences, 20(2) (2019-01-24)
Pipemidic acid (HPPA) is a quinolone antibacterial agent used mostly to treat gram-negative infections of the urinary tract, but its therapeutic use is limited because of its low solubility. Thus, to improve drug solubility, natural cyclodextrins (CDs) are used for
D A Schmid et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 36(1), 59-69 (2006-01-06)
Quinolones are widely used, broad spectrum antibiotics that can induce immediate- and delayed-type hypersensitivity reactions, presumably either IgE or T cell mediated, in about 2-3% of treated patients. To better understand how T cells interact with quinolones, we analysed six
Juan Li et al.
Environmental health perspectives, 120(8), 1144-1149 (2012-05-10)
Inappropriate use of antibiotics in swine feed could cause accelerated emergence of antibiotic resistance genes, and agricultural application of swine waste could spread antibiotic resistance genes to the surrounding environment. We investigated the distribution of plasmid-mediated quinolone resistance (PMQR) genes
I Grabnar et al.
International journal of pharmaceutics, 322(1-2), 52-59 (2006-06-30)
Intravesical administration of cytotoxic agents is commonly used in urological practice for treatment of superficial bladder cancer. The leading motive is optimisation of drug delivery near the site of action and reduction of systemic toxicity. Bladder pharmacokinetics is complicated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service