Skip to Content
Merck
All Photos(1)

Key Documents

P7623

Sigma-Aldrich

PKD2 Active human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), aqueous glycerol solution

Synonym(s):

Protein Kinase D2

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$648.00

$648.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 μG
$648.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$648.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥70% (SDS-PAGE)

form

aqueous glycerol solution

specific activity

100 units/mg protein (Bradford)

mol wt

protein apparent mol wt ~140 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKD2(25865)

Application

Kinase activity is measured as the molar amount of phosphate incorporated into the CREBTIDE substrate peptide (KRREILSRRPSYR), per minute per mg protein at 30 °C using a final concentration of 50 μM [32P] ATP.

Biochem/physiol Actions

PKD2 is a novel phorbol ester- and growth factor-stimulated serine/threonine kinase that contains two cysteine-rich motifs at the N terminus, a pleckstrin homology domain, and a catalytic domain. The PKD family of enzymes have been implicated in very diverse cellular functions, including Golgi organization and plasma membrane directed transport, metastasis, immune responses, apoptosis and cell proliferation.

Unit Definition

One unit will incorporate one nanomole of phosphate into the CREBTIDE substrate (KRREILSRRPSYR) per minute at 30 °C at pH 7.2 using a final concentration of 50 μM [32P] ATP.

Physical form

Solution of 50 μl in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM dithiothreitol (DTT), 0.1 mM EGTA, and 30% glycerol.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service