Skip to Content
Merck
All Photos(1)

Key Documents

P7568

Sigma-Aldrich

β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin

≥99%, lyophilized powder

Synonym(s):

1-O-Alkyl-2-acetyl-sn-glycero-3-phosphocholine, 2-Acetyl-1-alkyl-sn-glycero-3-phosphocholine, AcGEPC, PAF, Platelet activating factor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$244.00
5 MG
$718.00

$244.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$244.00
5 MG
$718.00

About This Item

CAS Number:
Beilstein:
4220745
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

$244.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

lyophilized powder

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C26H54NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5/h26H,6-24H2,1-5H3/t26-/m1/s1

InChI key

HVAUUPRFYPCOCA-AREMUKBSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phospholipid produced by lymphocytes, platelets and endothelial cells that induces platelet aggregation and is involved in inflammation, anaphylaxis and wound repair.

Packaging

Packaged under argon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Bussolino et al.
Cell calcium, 5(5), 463-473 (1984-10-01)
Platelet-activating factor (1-0-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine, PAF) is a potent inducer of shape-change, aggregation and secretion in platelets. PAF causes a rapid increase in intracellular calcium, but has no calcium gating effect in intact lipid bilayers. Human red cells (RBC) did not metabolize
Julie R Jin et al.
Frontiers in immunology, 11, 585070-585070 (2020-11-17)
Mast cells are the major effector cells in immunoglobulin E (IgE)-mediated allergy. The high affinity IgE receptor FcεRI, as well as G protein-coupled receptors (GPCRs) on the mast cell surface signals to phosphoinositide 3-kinase γ (PI3Kγ) to initiate degranulation, cytokine
Stephanie Saul et al.
Journal of leukocyte biology, 106(5), 1101-1115 (2019-06-20)
Neutrophils play a central role in immunity and inflammation via their intrinsic ability to migrate into inflamed tissue, to phagocytose pathogens, and to kill bacterial and fungi by releasing large quantities of superoxide anions and lytic enzymes. The molecular pathways

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service