Skip to Content
Merck
All Photos(1)

Key Documents

P6335

Supelco

Pramoxine hydrochloride

BioXtra

Synonym(s):

4-[3-(4-Butoxyphenoxy)propyl]morpholine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3 · HCl
CAS Number:
Molecular Weight:
329.86
EC Number:
MDL number:
UNSPSC Code:
41116107

product line

BioXtra

impurities

<0.0005% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

solubility

H2O: 0.5 M at 20 °C, clear, faintly yellow

anion traces

sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

SMILES string

Cl.CCCCOc1ccc(OCCCN2CCOCC2)cc1

InChI

1S/C17H27NO3.ClH/c1-2-3-12-20-16-5-7-17(8-6-16)21-13-4-9-18-10-14-19-15-11-18;/h5-8H,2-4,9-15H2,1H3;1H

InChI key

SYCBXBCPLUFJID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Topical anesthetic for skin or mucous membranes

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service