Key Documents
P6300
Pipet-Aid® automatic pipette pumps
cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
plastic
silicone
feature
CE compliant
manufacturer/tradename
Drummond Scientific 4-000-200
volume
1-100 mL
General description
Safe, convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.
Legal Information
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)