Skip to Content
Merck
All Photos(1)

Key Documents

P6278

Sigma-Aldrich

Peroxidase Inactivated

≤0.02 units/mg protein (biuret)

Synonym(s):

Peroxidase, inactivated from horseradish peroxidase

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$557.00
100 MG
$2,650.00

$557.00


Estimated to ship onMay 17, 2025



Select a Size

Change View
10 MG
$557.00
100 MG
$2,650.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$557.00


Estimated to ship onMay 17, 2025


form

powder

Quality Level

200
300

specific activity

≤0.02 units/mg protein (biuret)

impurities

salt, free

solubility

0.85% sodium chloride: soluble 15 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

2-8°C

Application

Inactivated peroxidase is used to study regeneration of inactivated peroxidase enzymes[1]. It has been used to study the mechanism of peroxidase solubilization[2] and it is used to study antitumor activity of immobilized peroxidase systems[3]. It may be used for developing differential staining techniques in nerve tissue preparations.

Biochem/physiol Actions

HRP readily combines with hydrogen peroxide (H2O2) and the resultant [HRP-H2O2] complex can oxidize a wide variety of hydrogen donors. The optimal pH is 6.0-6.5 and the enzyme is most stable in the pH range of 5.0-9.0. HRP can be conjugated to antibodies by several different methods that include the use of glutaraldehyde, periodate oxidation, disulfide bonds, and also via amino- and thiol-directed cross-linkers. It is smaller and more stable than the enzyme labels, β-galactosidase and alkaline phosphatase. Hence, it is the most desired label. Also, its glycosylation leads to lower non-specific binding.[4] It is also used for the determination of glucose[5] and peroxides[6] in solution. Sodium azide, cyanide, L-cystine, dichromate, ethylenethiourea, hydroxylamine, sulfide, vanadate, p-aminobenzoic acid, and ions such as Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+, and Pb2+ are found to inhibit the enzyme activity.[7]
Inactivated peroxidase reacts with goat and rabbit anti-peroxidase. It may be reactivated various methods such as heating[1].

Unit Definition

One unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20°C.

Preparation Note

Prepared from Type VI horseradish peroxidase by a modification of the method of Teale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Forehand et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 2(5), 654-652 (1982-05-01)
This study of spinal cord injury in bullfrog (Rana catesbeiana) tadpoles using the neuroanatomical tracer horseradish peroxidase (HRP) was undertaken to determine (1) whether the same anatomical regions that normally give rise to ascending or descending spinal tracts do so
Dual projections of single neurons are visualized simultaneously: use of enzymatically inactive [3H]HRP.
N L Hayes et al.
Brain research, 165(2), 321-326 (1979-04-13)
L L Vacca et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 23(3), 208-215 (1975-03-01)
Immunoperoxidase techniques are presented which can be used to localize horseradish peroxidase-tracer in paraffin-embedded tissues of the central nervous system. Compared to histochemical methods using frozen sections, these immunologic techniques allow the use of stored, serial paraffin sections, and appear
S Morimoto et al.
The Journal of biological chemistry, 274(37), 26192-26198 (1999-09-03)
Elicitor treatment or mechanical damage to Scutellaria baicalensis Georgi (skullcap plants) callus causes an immediate insolubilization of a 36-kDa protein into cell walls. The 36-kDa protein was identified as peroxidase 1 by analysis of its internal amino acid sequence and
Antitumor Activity of an Immobilized Peroxidase System Against Murine Ehrlich Ascites Is Mediated by the Immune System
Steven H. Combs, Clinton D. Morgan, et al.
Exp. Biol. Med, 204, 306-311 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service