Skip to Content
Merck
All Photos(1)

Key Documents

P6210

Sigma-Aldrich

Pyrantel pamoate

BioXtra

Synonym(s):

1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Pyrantel embonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H14N2S · C23H16O6
CAS Number:
Molecular Weight:
594.68
Beilstein:
9034755
EC Number:
MDL number:
UNSPSC Code:
51452307
PubChem Substance ID:
NACRES:
NA.85

product line

BioXtra

form

powder

impurities

<0.0005% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.5%

cation traces

Al: <0.0005%
Ca: <0.002%
Cu: <0.0005%
Fe: <0.001%
K: <0.005%
Mg: <0.0005%
Na: <0.01%
Pb: <0.001%
Zn: <0.0005%

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

SMILES string

CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(O)=O)c3O

InChI

1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+

InChI key

AQXXZDYPVDOQEE-MXDQRGINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

It is a combination of pyrantel and pamoic acid. It is used to study mechanisms underlying gut dysfunction in a murine model.

Biochem/physiol Actions

Pyrantel pamoate is a nematode muscle cell-associated nicotinic agonist for the acetylcholine (ACh) receptor. Cholinesterase inhibitor.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kashinath Ghosh et al.
The Journal of infectious diseases, 193(1), 155-162 (2005-12-03)
The effect of concurrent (active) and treated hookworm infections on the immunogenicity of vaccination with the recombinant fusion protein Ay-Ancylostoma-secreted protein 2 was analyzed in the Golden Syrian hamster. Hamsters were infected with the hookworm Ancylostoma ceylanicum and vaccinated with
Education and imaging. Gastrointestinal: ascariasis.
A Suzuki et al.
Journal of gastroenterology and hepatology, 23(11), 1770-1770 (2009-01-06)
Fabrizia Veronesi et al.
Veterinary parasitology, 172(1-2), 164-167 (2010-05-18)
A study was carried out to assess the field efficacy of moxidectin (MOX) against Parascaris equorum in foals. A total of 70 foals from 2 Italian thoroughbred (Farms 1 and 2) stud farms and one trotter stud farm (Farm 3)
Vinod K Gupta et al.
Journal of colloid and interface science, 350(1), 330-335 (2010-07-21)
Adsorption and electrochemical reduction of pyrantel pamoate are studied in Britton Robinson buffer medium at hanging mercury drop electrode (HMDE) by Adsorptive Stripping Voltammetric technique. The peak current shows a linear dependence with the drug concentration over the range 250
C A Northrop-Clewes et al.
The American journal of clinical nutrition, 73(1), 53-60 (2000-12-22)
The effect of helminth infestation on the nutrition, growth, and physiology of the host is still poorly understood. Anthelmintic treatment of children in developing countries has had varying success in terms of growth improvements. The objective of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service