Skip to Content
Merck
All Photos(1)

Key Documents

P4618

Sigma-Aldrich

Anti-PUMA/bbc3, C-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Bcl-2 Binding Component 3, Anti-p53 Upregulated Modulator of Apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
conjugate:
unconjugated
application:
ARR
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
4
technique(s):
microarray: suitable
western blot: 1-2 μg/mL

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

microarray: suitable
western blot: 1-2 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BBC3(27113)
mouse ... Bbc3(170770)

Specificity

Using K562 or 3T3 cell lysates, the antibody recognizes a band at ~23 kDa. A lower band at ~16 kDa may be detected in MOLT4 and U937 cells, which may represent PUMA-β.

Immunogen

synthetic peptide corresponding to amino acids 180-193 of human PUMA-α.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Gangoda et al.
Cell death and differentiation, 21(11), 1815-1824 (2014-07-12)
Loss of function mutations in the Prkar1a gene are the cause of most cases of Carney complex disorder. Defects in Prkar1a are thought to cause hyper-activation of PKA signalling, which drives neoplastic transformation, and Prkar1a is therefore considered to be
Mustak Pirbhai et al.
The Journal of biological chemistry, 281(42), 31495-31501 (2006-08-31)
Chlamydia trachomatis has evolved a profound anti-apoptotic activity that may aid in chlamydial evasion of host defense. The C. trachomatis anti-apoptotic activity has been correlated with blockade of mitochondrial cytochrome c release, inhibition of Bax and Bak activation, and degradation
Jennifer Adamski et al.
PloS one, 8(6), e65304-e65304 (2013-06-21)
Survival rates from childhood cancer have improved dramatically in the last 40 years, such that over 80% of children are now cured. However in certain subgroups, including metastatic osteosarcoma, survival has remained stubbornly poor, despite dose intensive multi-agent chemotherapy regimens
Baharan Fekry et al.
Nature communications, 9(1), 4149-4149 (2018-10-10)
Ceramides are important participants of signal transduction, regulating fundamental cellular processes. Here we report the mechanism for activation of p53 tumor suppressor by C16-ceramide. C16-ceramide tightly binds within the p53 DNA-binding domain (Kd ~ 60 nM), in close vicinity to the Box V

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service