Skip to Content
Merck
All Photos(1)

Documents

P4153

Sigma-Aldrich

Prednisolone 21-hemisuccinate sodium salt

≥90%, powder

Synonym(s):

Δ1-Hydrocortisone 21-hemisuccinate, 1,4-Pregnadiene-11β,17,21-triol-3,20-dione 21-hemisuccinate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H31O8Na
CAS Number:
Molecular Weight:
482.50
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

sterility

non-sterile

Assay

≥90%

form

powder

color

white

solubility

water: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(=O)CCC(O)=O

InChI

1S/C25H32O8.Na.H/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;;/h7,9,11,16-18,22,27,32H,3-6,8,10,12-13H2,1-2H3,(H,29,30);;/t16-,17-,18-,22+,23-,24-,25-;;/m0../s1

InChI key

HLHQSPQPHMOTMP-FKIWUUNWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Shin-ichiro Kitajiri et al.
Archives of otolaryngology--head & neck surgery, 128(4), 365-367 (2002-04-03)
To assess the efficacy of corticosteroid therapy for sudden-onset sensorineural hearing loss at lower frequencies. Retrospective, nonrandomized, controlled study. The recovery rate, recovery/improvement rate, and period between the start of therapy and improvement of symptoms were studied in 2 groups:
Hong Yan et al.
Biological chemistry, 384(8), 1185-1194 (2003-09-17)
Previously we showed that glycation-induced inactivation and loss of antigenicity of enzymes occur simultaneously. Alpha-crystallin, a major structural protein of the mammalian lens, prevents the aggregation of other proteins and protects enzyme function against post-translational modification in vitro. However, it
Y Fujita et al.
Orthodontics & craniofacial research, 11(4), 187-195 (2008-10-28)
To analyse the effects of short-term treatment with etidronate on the glucocorticoid-induced retardation of bone growth and deterioration of bone structure in the prepubertal rat mandible. Fifty 5-week-old male rats were divided into five groups. Etidronate or vehicle treatment (5
Joanna Folwarczna et al.
Pharmacological reports : PR, 63(4), 1040-1049 (2011-10-18)
Glucocorticoid-induced osteoporosis is the most frequently occurring type of secondary osteoporosis. Antagonists of β-adrenergic receptors are now considered to be potential drugs under investigation for osteoporosis. The aim of the present study was to investigate the effects of propranolol, a
Toru Ohashi et al.
Acta oto-laryngologica, 132(2), 133-140 (2011-12-29)
Abnormal action potential (AP) recovery from adaptation was manifested as increased recovery (iR) or decreased recovery (dR) relative to the normal recovery range. dR was not analyzed statistically due to the very small number of cases. iR is considered an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service