Skip to Content
Merck
All Photos(2)

Key Documents

P3988

Sigma-Aldrich

20S Proteasome Fraction from rabbit

≥95% (SDS-PAGE), solution

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$935.00

$935.00


Estimated to ship onMay 27, 2025



Select a Size

Change View
25 μG
$935.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$935.00


Estimated to ship onMay 27, 2025


biological source

rabbit

Quality Level

Assay

≥95% (SDS-PAGE)

form

solution

mol wt

700 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Manufactured for Sigma by Boston Biochem., Inc.

Application

20S proteasome fraction from rabbit has been used as a positive control to determine proteasome activity.[1]

Biochem/physiol Actions

Catalytic core of the 26S proteasome that degrades polyubiquitinated proteins.
Hydrolyzes various peptide substrates and proteins with broad specificity in a non-ATP dependent process.

Physical form

Solution in 50 mM HEPES, pH 7.6, 150 mM sodium chloride, and 1 mM DTT, pH 7.6.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hough et al.
The Journal of biological chemistry, 262(17), 8303-8313 (1987-06-15)
We have purified two high molecular weight proteases approximately 400-fold from rabbit reticulocyte lysate. Both enzymes hydrolyze 125I-alpha-casein and 4-methylcoumaryl-7-amide peptides with tyrosine, phenylalanine, or arginine at the P1 position. Both are inhibited by hemin, thiol reagents, chymostatin, and leupeptin.
Examination of `lipotoxicity? in skeletal muscle of high-fat fed and ob/ob mice
S.M. Turpin
The Journal of Physiology (2009)
R L Stein et al.
Biochemistry, 35(13), 3899-3908 (1996-04-02)
In this paper, we report kinetic studies for the chymotryptic activity of the 20S proteasome. Major observations include the following: (1) Reaction progress curves that are recorded at concentrations of Suc-Leu-Leu-Val-Tyr-AMC greater than about 40 microM are biphasic and characterized
S M Turpin et al.
The Journal of physiology, 587(Pt 7), 1593-1605 (2009-02-11)
Excess lipid accumulation resulting from an elevated supply of plasma fatty acids is linked to the pathogenesis of the metabolic syndrome and heart disease. The term 'lipotoxicity' was coined to describe how lipid accumulation leads to cellular dysfunction and death

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service