Skip to Content
Merck
All Photos(1)

Key Documents

P3871

Sigma-Aldrich

pT7-FLAG-SBP-1 Expression Vector

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$417.00

$417.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$417.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.84

$417.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

shipped in

dry ice

storage temp.

−20°C

General description

Cytoplasmic expression of Met-N-terminal FLAG®, C-terminal SBP (Steptavidin Binding Peptide) dual tagged fusion proteins under the T7/lac promoter.

Other Notes

Supplied with pT7-SBP-1+BAP Control Vector.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service