Skip to Content
Merck
All Photos(1)

Key Documents

P3496

Sigma-Aldrich

(+/-)-8-Prenylnaringenin

Plant-derived estrogen receptor ligand

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$496.00

$496.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C20H20O5
Molecular Weight:
340.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.56

$496.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

storage temp.

−20°C

SMILES string

C\C(C)=C\Cc1c(O)cc(O)c2C(=O)CC(Oc12)c3ccc(O)cc3

InChI

1S/C20H20O5/c1-11(2)3-8-14-15(22)9-16(23)19-17(24)10-18(25-20(14)19)12-4-6-13(21)7-5-12/h3-7,9,18,21-23H,8,10H2,1-2H3

InChI key

LPEPZZAVFJPLNZ-UHFFFAOYSA-N

Biochem/physiol Actions

8-Prenylnaringenin, a strong plant-derived estrogen receptor ligand, is a phytoestrogen that inhibits multidrug resistance-associated transporters, P-glycoprotein and MRP1. 8-Prenylnaringenin also inhibits EGF signaling at the level of phosphatidylinositol-3-OH kinase ((PI(3)K))activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Sehmisch et al.
Planta medica, 74(8), 794-801 (2008-06-10)
As the average age of society increases, identifying and preventing osteoporosis becomes more important. According to the results of the Women's Health Initiative study, substitution of estradiol is not recommended in hormone replacement therapy (HRT), although phytoestrogens might be a
Elisa Brunelli et al.
The Journal of steroid biochemistry and molecular biology, 113(3-5), 163-170 (2008-12-24)
8-Prenylnaringenin (8PN), one of the strongest plant-derived oestrogen receptors (ERs) ligand, has been suggested to have potential cancer chemo-preventive activities and anti-angiogenic properties. Because published data suggest that ERs serve as nodal point that allows interactions between hormones and growth
Olga Wesołowska et al.
European journal of pharmacology, 644(1-3), 32-40 (2010-07-17)
Flavonoids with hydrophobic e.g. prenyl substituents might constitute the promising candidates for multidrug resistance (MDR) reversal agents. The interaction of 8-prenylnaringenin (8-isopentenylnaringenin), a potent phytoestrogen isolated from common hop (Humulus lupulus), with two multidrug resistance-associated ABC transporters of cancer cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service