Skip to Content
Merck
All Photos(3)

Key Documents

P3405

Sigma-Aldrich

POPSO hydrate

≥99% (titration)

Synonym(s):

Piperazine-1,4-bis(2-hydroxypropanesulfonic acid) dihydrate, Piperazine-N,N′-bis(2-hydroxypropanesulfonic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$221.00

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$221.00

About This Item

Empirical Formula (Hill Notation):
C10H22N2O8S2 · xH2O
CAS Number:
Molecular Weight:
362.42 (anhydrous basis)
Beilstein:
842252
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99% (titration)

form

crystalline powder

useful pH range

7.2-8.5

pKa (25 °C)

7.8

solubility

1 M NaOH: 0.25 g/mL, clear to slightly hazy, colorless

application(s)

diagnostic assay manufacturing

SMILES string

O.OC(CN1CCN(CC1)CC(O)CS(O)(=O)=O)CS(O)(=O)=O

InChI

1S/C10H22N2O8S2.H2O/c13-9(7-21(15,16)17)5-11-1-2-12(4-3-11)6-10(14)8-22(18,19)20;/h9-10,13-14H,1-8H2,(H,15,16,17)(H,18,19,20);1H2

InChI key

VPTYCVRYPYNQLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service