Skip to Content
Merck
All Photos(3)

Key Documents

P3275

Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine

synthetic, ≥98%

Synonym(s):

1,2-Dihexadecanoyl-rac-glycero-3-phosphoethanolamine, rac-Phosphatidylethanolamine, 1,2-dipalmitoyl, DL-β,γ-Dipalmitoyl-α-cephalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96
Beilstein:
1730602
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥98%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)

InChI key

SLKDGVPOSSLUAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sathiah Thennarasu et al.
Biochemistry, 49(50), 10595-10605 (2010-11-11)
In a minimalist design approach, a synthetic peptide MSI-367 [(KFAKKFA)(3)-NH(2)] was designed and synthesized with the objective of generating cell-selective nonlytic peptides, which have a significant bearing on cell targeting. The peptide exhibited potent activity against both bacteria and fungi
Christopher F Monson et al.
Analytical chemistry, 83(6), 2090-2096 (2011-02-16)
A pH controlled flow cell device was constructed to allow electrophoretic movement of charged lipids and membrane associated proteins in supported phospholipid bilayers. The device isolated electrolysis products near the electrodes from the electrophoresis process within the bilayer. This allowed
Hui-Ling Sung et al.
Molecules (Basel, Switzerland), 17(7), 8533-8553 (2012-07-19)
This study presents a new reaction of cationic vinylidene complexes with Me₃SiN₃ (TMSN₃), which yields N-coordinated nitrile complexes 3. Treatment of a ruthenium acetylide precursor containing indenyl and dppe ligands with a series of organic halides produced the corresponding vinylidene
Michael S Jablin et al.
Biophysical journal, 101(1), 128-133 (2011-07-05)
Temperature-sensitive hydrogel polymers are utilized as responsive layers in various applications. Although the polymer's native characteristics have been studied extensively, details concerning its properties during interaction with biorelated structures are lacking. This work investigates the interaction between a thermoresponsive polymer
P J N Kett et al.
The journal of physical chemistry. B, 115(20), 6465-6473 (2011-05-06)
The structure of hybrid bilayer membranes (HBMs) containing either a pure cholesterol or mixed cholesterol/dipalmitoylphosphatidylethanolamine (DPPE) proximal layer adsorbed onto an octadecanethiol (ODT) self-assembled monolayer (SAM) on a gold substrate have been investigated by sum frequency generation (SFG) spectroscopy. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service