Skip to Content
Merck
All Photos(1)

Documents

P2959

Sigma-Aldrich

PD 142,893

≥97% (HPLC)

Synonym(s):

N-Acetyl-β-Phenyl-D-Phe-Leu-Asp-Ile-Ile-Trp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H65N7O10
CAS Number:
Molecular Weight:
924.09
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](NC(C)=O)C(c1ccccc1)c2ccccc2)[C@@H](C)CC)C(=O)N[C@@H](Cc3c[nH]c4ccccc34)C(O)=O

Amino Acid Sequence

N-Acetyl-β-Phenyl-Phe-Leu-Asp-Ile-Ile-Trp

General description

Contains the conserved endothelin sequence, His-Leu-Asp-Ile-Ile-Trp, with His replaced by a bulky aromatic moiety.

Biochem/physiol Actions

Non-selective endothelin receptor antagonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Drimal et al.
European journal of pharmacology, 396(1), 19-22 (2000-05-24)
The characteristics of specific binding of human [125I]Tyr(13)-endothelin-(1-21), [125I]-Tyr(13)-Suc-[Glu(9),Ala(11, 15)]-endothelin-(8-21), ([125I]IRL-1620) and endothelin ET(A) receptor antagonist [125I]Tyr(3)-(N-[(hexahydro-1H-azepin-1-yl)carbonyl]-L-Leu]-1Me )-D-Trp ([125I]PD151242) (number of sites and their affinity) and proliferation responses to exogenous endothelin receptor agonists (endothelin-1 and the endothelin ET(B) receptor-selective, truncated
T D Warner et al.
British journal of pharmacology, 110(2), 777-782 (1993-10-01)
1. We have compared the receptors mediating the contractions of rings of rat thoracic aorta or rabbit pulmonary artery and rat stomach strips in response to the endothelin/sarafotoxin (ET/SX) family of peptides and to those mediating endothelium-dependent vasodilations within the
R L Schroeder et al.
Journal of cardiovascular pharmacology, 32(6), 935-943 (1998-12-30)
The purpose of this study was to determine whether vascular endothelial and vascular smooth-muscle endothelin ET(B) receptors could be quantitatively differentiated by PD 142893 (PD), SB 209670 (SB), and BQ 788 (BQ) in the same species by using closely matched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service