Skip to Content
Merck
All Photos(1)

Key Documents

P2688

Millipore

Immobilon®-P Polyvinylidene difluoride membranes

size 20 cm × 20 cm

Synonym(s):

PVDF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

Pricing and availability is not currently available.

manufacturer/tradename

Millipore IPVH20200

size

20 cm × 20 cm

Looking for similar products? Visit Product Comparison Guide

Application

Polyvinylidene difluoride membranes are popular membranes for immunoblotting (Western blotting), and sequencing of proteins. The basis of protein binding to PVDF is believed to be due to hydrophobic interactions. PVDF membranes for Western blotting are available in the most commonly used 0.45 μm pore size. PVDF membranes have a greater protein binding capacity, are more durable and more solvent resistant than nitrocellulose.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clare M Hamilton et al.
Infection and immunity, 77(6), 2488-2498 (2009-04-01)
Parasitic worms and molecules derived from them have powerful anti-inflammatory properties and are shown to have therapeutic effects on inflammatory diseases. The helminth Fasciola hepatica has been reported to suppress antigen-specific Th1 responses in concurrent bacterial infections, thus demonstrating its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service