Skip to Content
Merck
All Photos(1)

Key Documents

P2165

Sigma-Aldrich

Protein A–Biotin from Staphylococcus aureus

lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$294.00

$294.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
2 MG
$294.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$294.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

conjugate

biotin conjugate

Quality Level

form

lyophilized powder

composition

Protein, ≥70% biuret

extent of labeling

3.0-8.0 mol biotin per mol Protein A

storage temp.

2-8°C

General description

Protein A (SpA) is a 42kDa virulence factor produced by Staphylococcus aureus, and is composed of five highly homologous consecutive extracellular Ig-binding domains, namely domains E, D, A, B, and C. This protein is present in both secreted and membrane-associated forms.[1]

Specificity

Binds IgG only from most mammals, except rat, goat, and sheep

Biochem/physiol Actions

Protein A (SpA) causes B cells death in vitro, by binding to the Fc region of antibody and the Fab regions of the B-cell receptor, thus inhibiting its opsonophagocytosis. This protein is thought to be implicated in the evasion of host immune responses by Staphylococcus aureus, and thus, might be a candidate for vaccine development approaches to prevent severe S. aureus infections.[2] S. aureus is responsible for nosocomial infections as well as a variety of human infections, such as food poisoning, toxic shock syndrome, endocarditis, septicemia, skin infections, soft tissue infections, and bone infections. It is also responsible for bovine and ovine mastitis. SpA typing or DNA sequence analysis of the variable repeat region of protein A gene is useful in determining S. aureus outbreak strains from those strains that are epidemiologically insignificant.[3]

Physical form

Lyophilized powder containing sodium citrate

Preparation Note

Prepared from P 6031, coupled to biotin by an amide bond through an aminocaproyl spacer.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Balasch et al.
Journal of comparative pathology, 121(2), 139-148 (1999-07-16)
This report describes the experimental inoculation of conventional pigs with a tissue homogenate obtained from two pigs affected with postweaning multisystemic wasting syndrome (PMWS). Eight 2-month-old pigs were inoculated by the intranasal route, and two pigs were left as uninfected
Anil K Pal et al.
Biosensors, 10(10) (2020-09-30)
We report a novel self-referenced diffraction-based leaky waveguide (LW) comprising a thin (~2 µm) film of a photofunctionalisable hydrogel created by covalent attachment of a biotinylated photocleavable linker to chitosan. Streptavidin attached to the chitosan via the photocleavable linker was
spa typing method for discriminating among Staphylococcus aureus isolates: implications for use of a single marker to detect genetic micro- and macrovariation.
Koreen L et al
Journal of Clinical Microbiology, 42(2), 792-799 (2004)
Regina Stoltenburg et al.
International journal of molecular sciences, 19(2) (2018-03-03)
New, as yet undiscovered aptamers for Protein A were identified by applying next generation sequencing (NGS) to a previously selected aptamer pool. This pool was obtained in a classical SELEX (Systematic Evolution of Ligands by EXponential enrichment) experiment using the
Scott D Kobayashi et al.
mBio, 4(5), e00764-e00713 (2013-10-03)
Staphylococcus aureus is a prominent cause of human infections worldwide and is notorious for its ability to acquire resistance to antibiotics. Methicillin-resistant S. aureus (MRSA), in particular, is endemic in hospitals and is the most frequent cause of community-associated bacterial

Questions

  1. what is the purity of this product?

    1 answer
    1. The purity of this material is not determined. The concentration of Protein A and Biotin are reported individually in the Certificate of Analysis. Please see the link below to review a sample or lot-specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/p2165#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service