Skip to Content
Merck
All Photos(4)

Key Documents

P1937

Sigma-Aldrich

Phosphocreatine di(tris) salt

≥97% (enzymatic)

Synonym(s):

Creatine phosphate di(tris) salt, N-(Imino[phosphonoamino]methyl)-N-methylglycine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$188.00
1 G
$314.00
5 G
$1,330.00

$188.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$188.00
1 G
$314.00
5 G
$1,330.00

About This Item

Empirical Formula (Hill Notation):
C4H10N3O5P · 2C4H11NO3
CAS Number:
Molecular Weight:
453.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$188.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Product Name

Phosphocreatine di(tris) salt, ≥97% (enzymatic)

biological source

rabbit muscle

Assay

≥97% (enzymatic)

form

powder

color

white

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

cation traces

Na: ≤0.1%

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.NC(CO)(CO)CO.CN(CC(O)=O)C(=N)NP(O)(O)=O

InChI

1S/C4H10N3O5P.2C4H11NO3/c1-7(2-3(8)9)4(5)6-13(10,11)12;2*5-4(1-6,2-7)3-8/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);2*6-8H,1-3,5H2

InChI key

UPPBKCOGWJTQMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in systems where alkali metal ions are undesirable.

Biochem/physiol Actions

High-energy phosphate reservoir in vertebrate and some invertebrate muscle; provides phosphate for ADP-ATP conversion
High-energy phosphate reservoir in vertebrate and some invertebrate muscle; provides phosphate for ADP-ATP conversion. As a non-oxidative reservoir of high-energy phosphate, phosphocreatine protects against anoxic and ischemic damage to brain neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofía Muñoz et al.
Molecular cell, 74(4), 664-673 (2019-03-30)
Cohesin is a conserved, ring-shaped protein complex that topologically embraces DNA. Its central role in genome organization includes functions in sister chromatid cohesion, DNA repair, and transcriptional regulation. Cohesin loading onto chromosomes requires the Scc2-Scc4 cohesin loader, whose presence on
Alexandra Colin et al.
Current biology : CB, 28(16), 2647-2656 (2018-08-14)
Coordination between actin filaments and microtubules is critical to complete important steps during cell division. For instance, cytoplasmic actin filament dynamics play an active role in the off-center positioning of the spindle during metaphase I in mouse oocytes [1-3] or
Guixin Zhu et al.
Cell reports, 33(9), 108418-108418 (2020-12-03)
Neurodegenerative diseases are characterized by the formation and propagation of protein aggregates, especially amyloid fibrils. However, what normally suppresses protein misfolding and aggregation in metazoan cells remains incompletely understood. Here, we show that TRIM11, a member of the metazoan tripartite
Lou Beaulieu-Laroche et al.
Neuron, 103(2), 235-241 (2019-06-11)
Dendritic integration can expand the information-processing capabilities of neurons. However, the recruitment of active dendritic processing in vivo and its relationship to somatic activity remain poorly understood. Here, we use two-photon GCaMP6f imaging to simultaneously monitor dendritic and somatic compartments in
Trace Thome et al.
American journal of physiology. Cell physiology, 317(4), C701-C713 (2019-07-11)
Chronic kidney disease (CKD) leads to increased skeletal muscle fatigue, weakness, and atrophy. Previous work has implicated mitochondria within the skeletal muscle as a mediator of muscle dysfunction in CKD; however, the mechanisms underlying mitochondrial dysfunction in CKD are not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service